(1S)-(+)-Camphor-10-sulphonic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C0015-25G C0015-500G C0015-100G
TCIAC0015-25GEA 23.1 EUR
TCIAC0015-25G TCIAC0015-500G TCIAC0015-100G
(1S)-(+)-Camphor-10-sulphonic acid ≥98.0% (by titrimetric analysis)
(1S)-(+)-Camphor-10-sulphonic acid
Formule: C₁₀H₁₆O₄S
Kookpunt: 375 °C (1013 hPa)
Smeltpunt: 200°C
MDL Nummer: MFCD00064157
CAS nummer: 3144-16-9
EINECS: 221-554-1
UN: 3261
ADR: 8,III
Merck Index: 14,01734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b07c69f8dd1963b4d63f3af7f3469951296ca49&vid=5b07c69f8dd1963b4d63f3af7f3469951296ca49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +20.0 to +23.0 deg (C=5, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR