4-Chloorbenzaldehyde ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C0125-25G C0125-500G C0125-100G
TCIAC0125-25GEA 30.8 EUR
TCIAC0125-25G TCIAC0125-500G TCIAC0125-100G
4-Chloorbenzaldehyde ≥97.0% (by GC)
4-Chloorbenzaldehyde
Formule: C₇H₅ClO
Kookpunt: 214°C
Smeltpunt: 46 °C
Dichtheid: 1,196 g/cm³ (20 °C)
Vlampunt: 88 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003379
CAS nummer: 104-88-1
EINECS: 203-247-4
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef0b25c7330957aa2927d1c5510a987bf6627d37&vid=0960b5168f46f51a8c248f9eb04a0baac89a1560&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR