1-Chloro-2-methylpropane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C0207-25ML C0207-500ML
TCIAC0207-25MLEA 17.6 EUR
TCIAC0207-25ML TCIAC0207-500ML
1-Chloro-2-methylpropane ≥98.0% (by GC)
1-Chloro-2-methylpropane
Formule: C₄H₉Cl
Kookpunt: 70°C
Smeltpunt: –131 °C
Dichtheid: 0,8829 g/cm³ (20 °C)
Vlampunt: –6°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000954
CAS nummer: 513-36-0
EINECS: 208-157-9
UN: 1127
ADR: 3,II
Merck Index: 14,05138
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1aa2ca8d1d572a521e5defbed9fb32093aaba04&vid=802a4f71aa6062c0b509660a2e5fa6bcb3e248e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR