1-Chloor-2-propanol ≥70% (by GC) (contains ca. 25% 2-Chloro-1-propanol)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1-Chloropropan-2-ol, Propylene chlorohydrin, Chloropropanol

C0269-25ML C0269-500ML
TCIAC0269-25MLEA 25.3 EUR
TCIAC0269-25ML TCIAC0269-500ML
1-Chloor-2-propanol ≥70% (by GC) (contains ca. 25% 2-Chloro-1-propanol)
1-Chloor-2-propanol
Formule: C₃H₇ClO
MW: 94.54 g/mol
Kookpunt: 132°C
Vlampunt: 52°C
MDL Nummer: MFCD00004530
CAS nummer: 127-00-4
EINECS: 204-819-6
UN: 2611
ADR: 6.1 II
Merck Index: 14,07851

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 70.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR