4-Chloortolueen ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C0297-25G C0297-500G
TCIAC0297-25GEA 17.6 EUR
TCIAC0297-25G TCIAC0297-500G
4-Chloortolueen ≥98.0% (by GC)
4-Chloortolueen
Formule: C₇H₇Cl
Kookpunt: 162°C
Smeltpunt: 6…8 °C
Dichtheid: 1,07 g/cm³ (20 °C)
Vlampunt: 53°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000631
CAS nummer: 106-43-4
EINECS: 203-397-0
UN: 2238
ADR: 3,III
Merck Index: 14,02171
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fecef722b78e15235d9865989deec48fe01a335c&vid=61e08409d3c03bfcc0cf18b7ee559e7c357464fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR