Croton oil

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C0421-25ML C0421-5ML
TCIAC0421-25MLEA 173 EUR
TCIAC0421-25ML TCIAC0421-5ML
Croton oil
Croton oil
MDL Nummer: MFCD00130880
CAS nummer: 8001-28-3
UN: 2810
ADR: 6.1,III
Merck Index: 14,02598

Order Now

Specification Test Results

Appearance Orange to Amber to Dark red clear liquid
Iodine Value 100 to 120


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR