Allyl cyclohexanepropionate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C0485-25ML C0485-500ML
TCIAC0485-25MLEA 41.8 EUR
TCIAC0485-25ML TCIAC0485-500ML
Allyl cyclohexanepropionate ≥98.0% (by GC)
Allyl cyclohexanepropionate

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Cyclohexanepropionic Acid Allyl Ester, 3-Cyclohexylpropionic Acid Allyl Ester

Formule: C₁₂H₂₀O₂
Kookpunt: 91°C
Vlampunt: 126°C
MDL Nummer: MFCD00059557
CAS nummer: 2705-87-5
EINECS: 220-292-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9254211a63c907a95b8c349ca8b7616c882e8395&vid=9254211a63c907a95b8c349ca8b7616c882e8395&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR