2-Cyclohexylcyclohexanol cis- and trans-mixture ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C0498-25G
TCIAC0498-25GEA 145 EUR
TCIAC0498-25G
2-Cyclohexylcyclohexanol cis- and trans-mixture ≥98.0% (by GC)
2-Cyclohexylcyclohexanol
Formule: C₁₂H₂₂O
Kookpunt: 264°C
MDL Nummer: MFCD00021278
CAS nummer: 6531-86-8
EINECS: 229-430-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cc84a2c008e6f20bc6a00d6e67447febedecce2&vid=4c4eb25b51909ffcb1caf7caafc9e900ce96507f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR