Kongorot ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: CI 22120, Dinatrium-3,3'-[[1,1'-bifenyl]-4,4'-diylbis(azo)]bis(4-aminonaftaleen-1-sulfonaat)

C0550-25G
TCIAC0550-25GEA 92.4 EUR
TCIAC0550-25G
Kongorot ≥98.0% (by HPLC)
Kongorot
Formule: C₃₂H₂₂N₆Na₂O₆S₂
Smeltpunt: > 360 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004028
CAS nummer: 573-58-0
EINECS: 209-358-4
Merck Index: 14,02498
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27d7ed766f62f13e1b2c65580d41dd1e54238b4&vid=d27d7ed766f62f13e1b2c65580d41dd1e54238b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark red to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Absorbance min. 0.270 (10 mg/L, pH 5.0, 488.0 to 492.0 nm)
Solubility in hot Water transparency
Drying loss max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR