N-Carbobenzoxy-L-aspartic acid ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C0629-25G C0629-5G
TCIAC0629-25GEA 64.9 EUR
TCIAC0629-25G TCIAC0629-5G
N-Carbobenzoxy-L-aspartic acid ≥99.0% (by titrimetric analysis)
N-Carbobenzoxy-L-aspartic acid
Formule: C₁₂H₁₃NO₆
MDL Nummer: MFCD00002719
CAS nummer: 1152-61-0
EINECS: 214-568-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 117.0 to 122.0 deg-C
Specific rotation [a]20/D +9.0 to +10.0 deg (C=7, AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR