1,3-Cyclohexanedione ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C0682-25G C0682-250G
TCIAC0682-25GEA 27.5 EUR
TCIAC0682-25G TCIAC0682-250G
1,3-Cyclohexanedione ≥98.0% (by GC, titration analysis)
1,3-Cyclohexanedione
Formule: C₆H₈O₂
Kookpunt: 235 °C (1013 hPa)
Smeltpunt: 105 °C
Dichtheid: 1,25 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00001585
CAS nummer: 504-02-9
EINECS: 207-980-0
Merck Index: 14,03178
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7bef83303d232cf8943574e6c014247e0540a9f&vid=7dc23c922f28bbb9f1417597662c8ea45d14118d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 103.0 to 106.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR