Cholesterol oleyl carbonate ≥70% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C0691-10G C0691-25G
TCIAC0691-10GEA 44 EUR
TCIAC0691-10G TCIAC0691-25G
Cholesterol oleyl carbonate ≥70% (by HPLC)
Cholesterol oleyl carbonate
Formule: C₄₆H₈₀O₃
MDL Nummer: MFCD00003634
CAS nummer: 17110-51-9
EINECS: 241-179-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a725a35d596bf9ad9202b558da3d59c2427ce37e&vid=a725a35d596bf9ad9202b558da3d59c2427ce37e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) (CAD) min. 70.0 area%
Specific rotation [a]20/D -21.0 to -25.0 deg (C=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR