1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C0788-25G C0788-500G
TCIAC0788-25GEA 22 EUR
TCIAC0788-25G TCIAC0788-500G
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)
Formule: C₈H₁₂O₄
MDL Nummer: MFCD00001465
CAS nummer: 1076-97-7
EINECS: 214-068-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7890872c6c6ad11d8830189a4083482c5c6c91d&vid=39c06fdc903583351db17bb4a69df1ac8cf328bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystaline
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR