Ethyl-2-oxocyclohexanecarboxylate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C0913-25ML C0913-500ML C0913-100ML
TCIAC0913-25MLEA 47.3 EUR
TCIAC0913-25ML TCIAC0913-500ML TCIAC0913-100ML
Ethyl-2-oxocyclohexanecarboxylate ≥95.0% (by GC)
Ethyl-2-oxocyclohexanecarboxylate
Formule: C₉H₁₄O₃
Kookpunt: 160°C
Vlampunt: 85°C
MDL Nummer: MFCD00001631
CAS nummer: 1655-07-8
EINECS: 216-731-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62c15b30eefb8caf44b941b59da588029d37b2fd&vid=62c15b30eefb8caf44b941b59da588029d37b2fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Acid Value max. 5.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR