2-Ethylhexyl chloroformate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C0927-25ML C0927-500ML
TCIAC0927-25MLEA 18.7 EUR
TCIAC0927-25ML TCIAC0927-500ML
2-Ethylhexyl chloroformate ≥98.0% (by titrimetric analysis)
2-Ethylhexyl chloroformate
Formule: C₉H₁₇ClO₂
Kookpunt: 107°C
Vlampunt: 101°C
MDL Nummer: MFCD00000643
CAS nummer: 24468-13-1
EINECS: 246-278-9
UN: 2748
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37cc880f9f056e2f03338aa5adf50a4afb715064&vid=37cc880f9f056e2f03338aa5adf50a4afb715064&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (Anilin Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR