β-Chloropropiophenone ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C0995-25G
TCIAC0995-25GEA 61.6 EUR
TCIAC0995-25G
β-Chloropropiophenone ≥96.0% (by GC)
β-Chloropropiophenone
Formule: (C₆H₅)C(O)CH₂CH₂Cl
Kookpunt: 147°C
Smeltpunt: 48…49 °C
Dichtheid: 1,181 g/cm³ (20 °C)
MDL Nummer: MFCD00000990
CAS nummer: 936-59-4
EINECS: 213-317-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e35965471f5853b81c7cb7519788da49e83ddcab&vid=e35965471f5853b81c7cb7519788da49e83ddcab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 46.0 to 50.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR