4-Chlorothioanisole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide

C1156-10G C1156-25G
TCIAC1156-10GEA 42.9 EUR
TCIAC1156-10G TCIAC1156-25G
4-Chlorothioanisole ≥98.0% (by GC)
4-Chlorothioanisole
Formule: C₇H₇ClS
MW: 158.65 g/mol
Kookpunt: 224°C
MDL Nummer: MFCD00013643
CAS nummer: 123-09-1
EINECS: 204-600-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85638ff184eb9713b5e5572d90c42974766ca902&vid=85638ff184eb9713b5e5572d90c42974766ca902&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR