(2-Cyanoethyl)triethoxysilane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C1259-25ML C1259-250ML
TCIAC1259-25MLEA 29.7 EUR
TCIAC1259-25ML TCIAC1259-250ML
(2-Cyanoethyl)triethoxysilane ≥98.0% (by GC)
(2-Cyanoethyl)triethoxysilane
Formule: NCCH₂CH₂Si(OCH₂CH₃)₃
Kookpunt: 137°C
Dichtheid: 0,9792 g/cm³ (20 °C)
Vlampunt: 86°C
MDL Nummer: MFCD00019851
CAS nummer: 919-31-3
EINECS: 213-050-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a413f2d248aeaed6771d021f47938fae0c57cde9&vid=a413f2d248aeaed6771d021f47938fae0c57cde9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR