Benzyl chloroacetate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C1265-25G C1265-250G
TCIAC1265-25GEA 93.5 EUR
TCIAC1265-25G TCIAC1265-250G
Benzyl chloroacetate ≥97.0% (by GC)
Benzyl chloroacetate
Formule: C₉H₉ClO₂
Kookpunt: 132°C
Dichtheid: 1,2223 g/cm³ (4 °C)
MDL Nummer: MFCD00157062
CAS nummer: 140-18-1
EINECS: 205-400-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b2c76a691362060f8a53c5ecb434fa496253f2c&vid=7b2c76a691362060f8a53c5ecb434fa496253f2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR