Ethyl-2,4-dihydroxy-6-methylbenzoate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C1316-1G C1316-5G
TCIAC1316-1GEA 85.8 EUR
TCIAC1316-1G TCIAC1316-5G
Ethyl-2,4-dihydroxy-6-methylbenzoate ≥98.0% (by titrimetric analysis)
Ethyl-2,4-dihydroxy-6-methylbenzoate
Formule: C₁₀H₁₂O₄
MDL Nummer: MFCD00010821
CAS nummer: 2524-37-0

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR