α-Bromo-o-toluonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C1556-10G C1556-25G
TCIAC1556-10GEA 59.4 EUR
TCIAC1556-10G TCIAC1556-25G
α-Bromo-o-toluonitrile ≥98.0% (by GC)
α-Bromo-o-toluonitrile
Formule: C₈H₆BrN
Kookpunt: 111°C
Smeltpunt: 72…74 °C
MDL Nummer: MFCD00001794
CAS nummer: 22115-41-9
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e87c9cd8bb953159b061b115204431da0df0c1&vid=a0e87c9cd8bb953159b061b115204431da0df0c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 74.0 deg-C
Solubility in Methanol very faint turbidity

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR