4-Chloro-3-nitrobenzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C1712-25G C1712-5G
TCIAC1712-25GEA 186 EUR
TCIAC1712-25G TCIAC1712-5G
4-Chloro-3-nitrobenzyl alcohol ≥98.0% (by GC)
4-Chloro-3-nitrobenzyl alcohol
Formule: C₇H₆ClNO₃
Smeltpunt: 66 °C
MDL Nummer: MFCD00007086
CAS nummer: 55912-20-4
EINECS: 259-901-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9cf3d366e8c72759cc8f1f4cb72ec5bd7b319bf&vid=f9cf3d366e8c72759cc8f1f4cb72ec5bd7b319bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR