Chloro(diisopropyl)phosphine ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Di-i-propylchlorophosphine

C1720-5G
TCIAC1720-5GEA 134 EUR
TCIAC1720-5G
Chloro(diisopropyl)phosphine ≥98.0% (by titrimetric analysis)
Chloro(diisopropyl)phosphine
Formule: C₆H₁₄ClP
MW: 152.6 g/mol
Kookpunt: 158°C
Vlampunt: 4°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00015027
CAS nummer: 40244-90-4
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bfe44bb166ad52d9674835563ec022c117d47df&vid=2bfe44bb166ad52d9674835563ec022c117d47df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR