1-Chloroethylcyclohexyl carbonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C1786-25G C1786-250G
TCIAC1786-25GEA 25.3 EUR
TCIAC1786-25G TCIAC1786-250G
1-Chloroethylcyclohexyl carbonate ≥98.0% (by GC)
1-Chloroethylcyclohexyl carbonate
Formule: C₉H₁₅ClO₃
Vlampunt: 84°C
MDL Nummer: MFCD04038149
CAS nummer: 99464-83-2
EINECS: 444-950-8
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR