2-Chloro-4'-fluorobenzophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C1860-25G C1860-5G
TCIAC1860-25GEA 191 EUR
TCIAC1860-25G TCIAC1860-5G
2-Chloro-4'-fluorobenzophenone ≥98.0% (by GC)
2-Chloro-4'-fluorobenzophenone
Formule: C₁₃H₈ClFO
MDL Nummer: MFCD00000559
CAS nummer: 1806-23-1
EINECS: 217-300-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f78ee328be2036e18ed6371a8b93ed3f74beda9&vid=1f78ee328be2036e18ed6371a8b93ed3f74beda9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR