5-Chloro-2-hydroxy-3-nitropyridine ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C1972-25G C1972-5G
TCIAC1972-25GEA 279 EUR
TCIAC1972-25G TCIAC1972-5G
5-Chloro-2-hydroxy-3-nitropyridine ≥98.0% (by titrimetric analysis)
5-Chloro-2-hydroxy-3-nitropyridine
Formule: C₅H₃ClN₂O₃
MDL Nummer: MFCD00114884
CAS nummer: 21427-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=211462b000e35bf2b50c00f73a80007a3e991d3f&vid=211462b000e35bf2b50c00f73a80007a3e991d3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 93.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR