Cyclopentadienyltitanium trichloride ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C1994-1G C1994-5G
TCIAC1994-1GEA 50.6 EUR
TCIAC1994-1G TCIAC1994-5G
Cyclopentadienyltitanium trichloride ≥98.0% (by titrimetric analysis)
Cyclopentadienyltitanium trichloride
Formule: C₅Cl₃Ti
MDL Nummer: MFCD00003724
CAS nummer: 1270-98-0
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28af89522e0195d3fc2d4712e6eb2429c34dfa6c&vid=28af89522e0195d3fc2d4712e6eb2429c34dfa6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystaline
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR