2-Chloro-3-methyl-5-nitropyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C2020-1G C2020-5G
TCIAC2020-1GEA 26.4 EUR
TCIAC2020-1G TCIAC2020-5G
2-Chloro-3-methyl-5-nitropyridine ≥98.0% (by GC)
2-Chloro-3-methyl-5-nitropyridine
Formule: C₆H₅ClN₂O₂
Kookpunt: 145°C
MDL Nummer: MFCD00173686
CAS nummer: 22280-56-4
EINECS: 244-889-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=559a6514a11660f85a95577256aa269db6496cbc&vid=559a6514a11660f85a95577256aa269db6496cbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR