2-Chloro-5-fluorobenzonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C2080-1G C2080-5G
TCIAC2080-1GEA 31.9 EUR
TCIAC2080-1G TCIAC2080-5G
2-Chloro-5-fluorobenzonitrile ≥98.0% (by GC)
2-Chloro-5-fluorobenzonitrile
Formule: C₇H₃ClFN
MDL Nummer: MFCD03094168
CAS nummer: 57381-56-3
EINECS: 260-714-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b74ee43c8c01c573f20a833cd6df4eaeecf4bbd4&vid=b74ee43c8c01c573f20a833cd6df4eaeecf4bbd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR