Cyclohexyl-p-toluenesulphonate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C2363-25G C2363-5G
TCIAC2363-25GEA 132 EUR
TCIAC2363-25G TCIAC2363-5G
Cyclohexyl-p-toluenesulphonate ≥98.0% (by HPLC)
Cyclohexyl-p-toluenesulphonate
Formule: C₁₃H₁₈O₃S
MDL Nummer: MFCD00014291
CAS nummer: 953-91-3
EINECS: 213-468-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR