α-Bromo-3-chloro-5-fluorotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C2474-1G C2474-5G
TCIAC2474-1GEA 38.5 EUR
TCIAC2474-1G TCIAC2474-5G
α-Bromo-3-chloro-5-fluorotoluene ≥98.0% (by GC)
α-Bromo-3-chloro-5-fluorotoluene
Formule: C₇H₅BrClF
MDL Nummer: MFCD03788472
CAS nummer: 493024-39-8
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cca0bac6806a7defa41e92f52129e7b39ac0534d&vid=cca0bac6806a7defa41e92f52129e7b39ac0534d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR