4-Chloor-3-fluorfenol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C2476-25G C2476-5G
TCIAC2476-25GEA 297 EUR
TCIAC2476-25G TCIAC2476-5G
4-Chloor-3-fluorfenol ≥98.0% (by GC, titration analysis)
4-Chloor-3-fluorfenol
Formule: C₆H₄ClFO
Kookpunt: 84°C
Smeltpunt: 49…53 °C
MDL Nummer: MFCD00042583
CAS nummer: 348-60-7
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a113d9a8514215fab2ec0db383c28f7a09159f2e&vid=d92fa9a4289bcc4acddeda4fff42202ba40eaf12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR