N-Carbobenzoxy-D-2-phenylglycine ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (R)-2-(((Benzyloxy)carbonyl)amino)-2-phenylacetic acid, Z-D-Phg-OH, Z-D-2-phenylglycine, Cbz-D-Phg-OH

C2566-1G C2566-5G
TCIAC2566-1GEA 27.5 EUR
TCIAC2566-1G TCIAC2566-5G
N-Carbobenzoxy-D-2-phenylglycine ≥98.0% (by HPLC)
N-Carbobenzoxy-D-2-phenylglycine
Formule: C₁₆H₁₅NO₄
MW: 285.29 g/mol
MDL Nummer: MFCD00021703
CAS nummer: 17609-52-8
EINECS: 241-582-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 130.0 to 135.0 deg-C
Specific rotation [a]20/D -114.0 to -118.0 deg (C=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR