Clonixin ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C2619-25G
TCIAC2619-25GEA 94.6 EUR
TCIAC2619-25G
Clonixin ≥98.0% (by HPLC, titration analysis)
Clonixin
Formule: C₁₃H₁₁ClN₂O₂
MDL Nummer: MFCD00864292
CAS nummer: 17737-65-4
EINECS: 241-730-1
Merck Index: 14,02392

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 240.0 to 244.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR