Carmofur ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 5-Fluoro-N-hexyl-2,4-dioxo-3,4-dihydropyrimidine-1(2H)-carboxamide, 5-Fluoro-1-hexylcarbamoyluracil

C2663-5G
TCIAC2663-5GEA 872 EUR
TCIAC2663-5G
Carmofur ≥98.0% (by HPLC, titration analysis)
Carmofur
Formule: C₁₁H₁₆FN₃O₃
MW: 257.27 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00866284
CAS nummer: 61422-45-5
UN: 2811
ADR: 6.1 III
Merck Index: 14,01844

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR