(2-Chlorophenyl)acetylene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C2750-1G C2750-5G
TCIAC2750-1GEA 29.7 EUR
TCIAC2750-1G TCIAC2750-5G
(2-Chlorophenyl)acetylene ≥98.0% (by GC)
(2-Chlorophenyl)acetylene
Formule: C₈H₅Cl
Kookpunt: 72°C
Vlampunt: 70°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00269951
CAS nummer: 873-31-4

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR