7-Chloroisatin ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C2840-1G C2840-5G
TCIAC2840-1GEA 25.3 EUR
TCIAC2840-1G TCIAC2840-5G
7-Chloroisatin ≥98.0% (by GC, titration analysis)
7-Chloroisatin
Formule: C₈H₄ClNO₂
MDL Nummer: MFCD00022796
CAS nummer: 7477-63-6

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 187.0 to 191.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR