3-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C3279-1G C3279-5G
TCIAC3279-1GEA 84.7 EUR
TCIAC3279-1G TCIAC3279-5G
3-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-(4-methoxycarbonyl)phenylboronic acid
Formule: C₈H₈BClO₄
MDL Nummer: MFCD06659874
CAS nummer: 603122-82-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4f31663711f016efa68543506d0a59c464b5de0&vid=e4f31663711f016efa68543506d0a59c464b5de0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR