1-Cyclopentenylboronic acid pinacol ester ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C3385-1G C3385-5G
TCIAC3385-1GEA 36.3 EUR
TCIAC3385-1G TCIAC3385-5G
1-Cyclopentenylboronic acid pinacol ester ≥98.0% (by GC)
1-Cyclopentenylboronic acid pinacol ester
Formule: C₁₁H₁₉BO₂
MDL Nummer: MFCD08669391
CAS nummer: 287944-10-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR