4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C3555-1G C3555-5G
TCIAC3555-1GEA 49.5 EUR
TCIAC3555-1G TCIAC3555-5G
4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)
4-Cyano-3-fluorophenyl-4-n-pentylbenzoate
Formule: C₁₉H₁₈FNO₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD11053363
CAS nummer: 86786-89-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR