Cesium Lead tribromide, low water content ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

C3569-1G C3569-5G
TCIAC3569-1GEA 59.4 EUR
TCIAC3569-1G TCIAC3569-5G
Cesium Lead tribromide, low water content ≥98.0% (by titrimetric analysis)
Cesium Lead tribromide
Formule: Br₃CsPb
Smeltpunt: 555°C
CAS nummer: 15243-48-8
UN: 2291
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR