Cefoxitin sodium ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C3602-1G
TCIAC3602-1GEA 62.7 EUR
TCIAC3602-1G
Cefoxitin sodium ≥98.0% (by HPLC, titration analysis)
Cefoxitin sodium
Formule: C₁₆H₁₆N₃NaO₇S₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00079042
CAS nummer: 33564-30-6
EINECS: 251-574-6
Merck Index: 14,01936

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 202.0 to 206.0 deg-C
Specific rotation [a]20/D +206 to +214 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR