2-Cyano-2-propyl Dodecyl trithiocarbonate ≥97.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

C3627-1G
TCIAC3627-1GEA 118 EUR
TCIAC3627-1G
2-Cyano-2-propyl Dodecyl trithiocarbonate ≥97.0% (by HPLC, total nitrogen)
2-Cyano-2-propyl Dodecyl trithiocarbonate
Formule: C₁₇H₃₁NS₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD11973800
CAS nummer: 870196-83-1

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Specific gravity (20/20) 0.9950 to 1.0050


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR