1,4-Diaminobutane dihydrochloride ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D0081-25G D0081-500G
TCIAD0081-25GEA 46.2 EUR
TCIAD0081-25G TCIAD0081-500G
1,4-Diaminobutane dihydrochloride ≥98.0% (by titrimetric analysis)
1,4-Diaminobutane dihydrochloride
Formule: C₄H₁₄Cl₂N₂
Smeltpunt: 290°C
Dichtheid: 1,31 g/cm³ (20 °C)
MDL Nummer: MFCD00012526
CAS nummer: 333-93-7
EINECS: 206-375-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db36cdb25dff162482d60fe6d38c6d334f5bed53&vid=db36cdb25dff162482d60fe6d38c6d334f5bed53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR