1,5-Dibromopentane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D0198-25G D0198-500G D0198-100G
TCIAD0198-25GEA 18.7 EUR
TCIAD0198-25G TCIAD0198-500G TCIAD0198-100G
1,5-Dibromopentane ≥98.0% (by GC)
1,5-Dibromopentane
Formule: BrH₂C(CH₂)₃CH₂Br
Kookpunt: 221°C
Smeltpunt: –39 °C
Dichtheid: 1,7 g/cm³ (20 °C)
Vlampunt: 79°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000268
CAS nummer: 111-24-0
EINECS: 203-849-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c324c2f9ac10f7f475cd0491f91ee397f261803&vid=cb43642e15dc50aeaf766396e994cab610caf524&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR