α,α'-Dibromo-o-xylene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D0214-25G D0214-100G
TCIAD0214-25GEA 50.6 EUR
TCIAD0214-25G TCIAD0214-100G
α,α'-Dibromo-o-xylene ≥98.0% (by GC)
α,α'-Dibromo-o-xylene
Formule: C₈H₈Br₂
Kookpunt: 140°C
Smeltpunt: 90…93 °C
Dichtheid: 1,96 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000175
CAS nummer: 91-13-4
EINECS: 202-042-7
UN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec6ee5ea723bfefba680edea056a2ef73c65b42b&vid=ec6ee5ea723bfefba680edea056a2ef73c65b42b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 96.0 deg-C
Solubility in Toluene almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR