2,4-Dichlorobenzaldehyde ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D0330-25G D0330-500G
TCIAD0330-25GEA 17.6 EUR
TCIAD0330-25G TCIAD0330-500G
2,4-Dichlorobenzaldehyde ≥95.0% (by GC)
2,4-Dichlorobenzaldehyde
Formule: C₇H₄Cl₂O
Kookpunt: 233 °C (1013 hPa)
Smeltpunt: 68…71 °C
Dichtheid: 1,33 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003305
CAS nummer: 874-42-0
EINECS: 212-861-1
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5b71a599979e6e21bd8ab845d180ffe4cce2f5c&vid=4dda36c6d6e5bad9b48f02a1d0df03c0125c262a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Melting point 71.0 to 74.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR