2,6-Dichloorfenol ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D0394-25G D0394-500G
TCIAD0394-25GEA 62.7 EUR
TCIAD0394-25G TCIAD0394-500G
2,6-Dichloorfenol ≥99.0% (by GC)
2,6-Dichloorfenol
Formule: C₆H₄Cl₂O
MW: 163 g/mol
Kookpunt: 80°C
Smeltpunt: 68
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002176
CAS nummer: 87-65-0
EINECS: 201-761-3
UN: 2020
ADR: 6.1 III
Merck Index: 14,03073
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b19d92a090ed30412606957d88589da6ad7cfeb&vid=3bf6ff6aec19da63c4cb3d82cf2bba79f0b95f06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 65.0 to 68.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR