2-(2-Hexyloxyethoxy)ethanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D0501-25ML D0501-500ML
TCIAD0501-25MLEA 17.6 EUR
TCIAD0501-25ML TCIAD0501-500ML
2-(2-Hexyloxyethoxy)ethanol ≥98.0% (by GC)
Diethylene glycol monohexyl ether
Formule: C₁₀H₂₂O₃
Vlampunt: 141°C
MDL Nummer: MFCD00010703
CAS nummer: 112-59-4
EINECS: 203-988-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6449fb609dfe562c1c51ac42bbb1e68aa809f055&vid=6449fb609dfe562c1c51ac42bbb1e68aa809f055&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR