2,2'-Dibenzothiazole disulphide ≥96.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D0538-25G D0538-500G
TCIAD0538-25GEA 18.7 EUR
TCIAD0538-25G TCIAD0538-500G
2,2'-Dibenzothiazole disulphide ≥96.0% (by HPLC, total nitrogen)
2,2'-Dibenzothiazole disulphide
Formule: C₁₄H₈N₂S₄
MDL Nummer: MFCD00022874
CAS nummer: 120-78-5
EINECS: 204-424-9
UN: 3077
ADR: 9,III
Merck Index: 14,03370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77ca801d60e7b6c1f9d81c7c9fe584276c04eacb&vid=77ca801d60e7b6c1f9d81c7c9fe584276c04eacb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR